Learn More
Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
Brand:  Corning™ 99445-16XX
117.91 EUR valid until 2025-03-31
Use promo code "24073" to get your promotional price.
 View more versions of this product
Product Code. 10018522
  • 148.72 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed. Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
23 mL
Borosilicate Glass
For tissue culture work and general bacteriological use
Bulk
Non-sterile
16 mm
150 mm
Single-use
None
No
No
Culture Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Promotions
Promotions
  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073
    Expires2025-04-25
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    31
    Hours
    11
    Minutes
    23
    Seconds
    36
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes > 150 mm; 23 mL