missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific Chemicals

Product Code. p-7214723 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17231286

Brand: Thermo Scientific Acros 468990010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159326-68-8
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD08234647
InChI Key VSPXQZSDPSOPRO-UHFFFAOYSA-N
IUPAC Name pyrrolo[2,1-f][1,2,4]triazin-4-amine
SMILES NC1=NC=NN2C=CC=C12

Specifications

Quantity 1 g
Formula Weight 134.12
Chemical Name or Material Pyrrolo[2,1-f][1,2,4 ]triazin-4-amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.