missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Q-VD-OPH Inhibitor Peptide

Product Code. 18180784 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
This item is not returnable. View return policy

Product Code. 18180784

Brand: Novus Biologicals™ NBP2293915MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Functional, In vitro assay

TRUSTED_SUSTAINABILITY

Specifications

Host Species Human
Color Off-white
For Use With (Application) Functional, In vitro assay
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type Q-VD-OPH Inhibitor Peptide
Molecular Weight (g/mol) 513
Inhibitors Q-VD-OPH
Form Semi-solid

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.