Learn More
Thermo Scientific™ Quanti-Cult™ Candida albicans ATCC™ 10231™
Description
Thermo Scientific™ Quanti-Cult™ Candida albicans ATCC™ 10231™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum. 1 test/reconstituted vial; 10 tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
Specifications
Specifications
| ATCC® | 10231TM* |
| Description | Candida albicans ATCC™ 10231™ |
| Format | Quanti-Cult |
| Product Type | Bactidisk |
| Target Organism Class | Fungi |
| Test Type | Quantitative QC |
| Packaging Type | 10 Test/Kit |
| Strain Designations | 3148 [CBS 6431, CCY 29-3-106, CIP 48.72, DSM 1386, IFO 1594, NCPF 3179, NCYC 1363, NIH 3147, VTT C-85161] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.