Learn More
Quickfit™ Borosilicate Glass Flange Lid
Product Code. 15557920
Click to view available options
Description:
0° Side Socket: 14/23;, 5°, 10°, 15° Side Sockets: 19/26, Vertical Center Socket: 19/26
0°, 10° Side Sockets: 19/26, 5° Side Socket: 24/29, Vertical Center Socket: 19/26
0°, 10° and 2 x 5° Side Sockets: 19/26, Vertical Center Socket: 19/26
0°, 2 x 10° Side Sockets: 19/26, Vertical Center Socket: 34/35
0°, 5°, 10° Side Sockets: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 19/26
0°, 5°, 10° Side Sockets: 19/26, 15° Side Socket: 34/35, Vertical Center Socket:: 19/26
0°, 5°, 10° Side Sockets: 19/26, 15° Side Socket: 34/35, Vertical Center Socket:: 24/29
0°, 5°, 10°, Side Socket: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 29/32
Vertical Center Socket: 29/32
Vertical Center Socket: 34/35
Closure Size:
75 mm
100 mm
Unit Size:
Each
Pack of 5
Product Code. 15557920
Brand: Quickfit MAF2/41
Please to purchase this item. Need a web account? Register with us today!
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid includes precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid with one or more sockets to accept other Quickfit™ components.
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid is comprised of precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid includes one or more sockets to accept other Quickfit™ components.
  • With precision ground flat-flange joints
  • Fits Quickfit™ wide neck reaction flasks and culture vessels
  • Each lid with one or more sockets to accept other Quickfit™ components
TRUSTED_SUSTAINABILITY
Specifications
Description 0°, 5°, 10° Side Sockets: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 19/26
Material Borosilicate Glass
For Use With (Equipment) Wide neck flasks
Includes Sockets for use with other Quickfit™ components
Product Type Flange lid
Closure Size 100 mm