Learn More
Quickfit™ Borosilicate Glass Vigreux Fractionating Column
Product Code. p-8002936
Click to view available options
Height (Metric):
360 mm
600 mm
Unit Size:
Pack of 5
Product Code. 15527820
Brand: Quickfit FC3/14
Please to purchase this item. Need a web account? Register with us today!
Vigreux type with combination of horizontal and angled indentations.
  • Popular, easy to use column
  • Vigreux type with combination of horizontal and angled indentations
  • Horizontal indentations ensure intimate vapour and liquid contact
  • Angled indentations redistribute liquid from walls to center of column
TRUSTED_SUSTAINABILITY
Specifications
Includes 2 NS 29/32 standard ground joint
Height (Metric) 360 mm
Length (Metric) 360mm
Type Vigreux Column
Material Glass
Quantity 1Each
Standard Taper Joint NS 29/32