Learn More
Quickfit™ Stirrer Guides
Designed to reduce excessive whip in glass stirrer shafts which can lead to breakage.
Brand: Quickfit ST2/2/SC
This item is not returnable.
View return policy
Description
- PTFE bearing is fitted to the bottom of guides ST2/33C/SC and ST2/44C/SC for additional support
- Features a side-arm with detachable plastic connector for the introduction of gases or connection to a vacuum supply
Specifications
To reduce excessive 'whip' in Glass stirrer shafts which can lead to breakage | |
Tube connector, PTFE bearing, detachable Plastic connector | |
200 mm | |
19/26 |