Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
Brand: Quickfit ST54/18
44.27 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
- With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
- Ideal for securing thermometers, gas inlet tubes, etc
Specifications
29/32 | |
Plastic cap, Silicone Rubber ring and PTFE washer | |
Cone screw thread adapter |
Ideal for securing thermometers, gas inlet tubes | |
Borosilicate Glass | |
18 |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.