Learn More
Plum™ QuickFix Elastic Plaster Refill
Elastic and skin-sensitive textile plasters
Brand: Plum™ 5512
7.32 EUR valid until 2025-04-18
Use promo code "24095" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
6 x 45/Pk. | |
72 x 25 mm |
Promotions
-
Multiple Manufacturers
Outlet Corner
Use code 24095 to receive this promotion.Promo Code: 24095Expires: 2025-04-15
Days
12Hours
17Minutes
59Seconds
19
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Plum™ QuickFix Elastic Plaster Refill > Dimensions (L x W): 72 x 25 mm
Spot an opportunity for improvement?Share a Content Correction