Learn More
Sartorius Quintix™ Semi-Micro Balance with Internal Adjustment
The Quintix™ Semi-Micro Balance with Internal Adjustment makes your routine lab work easy with our revolutionary user interface.
Brand: Sartorius QUINTIX125D-1CEU
This item is not returnable.
View return policy
Description
- Redefining StandardQuintix™ makes laboratory work more enjoyable.
- The intuitive touch-screen in combination with precise weighing technology make Quintix™ laboratory balances a great choice for achieving the best results.
- You will benefit from improved workflows by easy-to-use application programs.
- If you need to generate GLP-compliant documents, you will save considerable time and effort with the ''plug and work'' connectivity of Quintix™ laboratory balances.
- Intuitive operation of the balance without having to read a manual thanks to the new user interface and color touch screen Integrated application programs and intuitive user guidance for greater efficiency in your daily lab routine
- Highest accuracy and convenience ensured at all times by the isoCAL fully automatic internal adjustment feature
- Easier record-keeping using the balance's directly integrated PC functions and Plug and Work connectivity to a Sartorius lab printer
- What's included:Universal power adapter, in-use cover, application guide, dust cover for analytical balances
Specifications
CFR | |
Touchscreen | |
≤4.0 sec., 1.5 sec. | |
218 mm |
60 to 120 g | |
0.01 mg, 0.1 mg | |
Balance |