missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Amino-3-phenylpropionic acid, 95%
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Quantity | 250 mg |
| Formula Weight | 165.19 |
| Sensitivity | Air Sensitive |
| Percent Purity | 95% |
| Chemical Name or Material | (R)-3-Amino-3-phenylpropionic acid |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu