Learn More
R&D Systems™ Human Axl (AAH32229) VersaClone cDNA
Axl Receptor Tyrosine Kinase
Brand:  R&D Systems™ RDC0598
Product Code. 18461390
  • 747.00 € / 10µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Human Axl (AAH32229) VersaClone cDNA clone contains the complete ORF for Axl, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Axl insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hAXL
10 μg at 0.2 μg/μL
2698bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
AAH32229
cDNA is provided in 10mM Tris-Cl, pH 8.5
AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Reasearch Use Only; Not for Use in Humans