missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein
CHO derived
Brand: R&D Systems™ 338-AC-01M/CF
This item is not returnable.
View return policy
Description
Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.Specifications
P08476 | |
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. | |
M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer) | |
1 mg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100-500 μg/mL in sterile 4 mM HCl. | |
Activin A |
Bioactivity | |
3624 | |
Activin A | |
Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426 | |
Disulfide-linked homodimer | |
INHBA | |
Recombinant | |
>95%, by SDS-PAGE with silver staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction