Learn More
R&D Systems™ Human MRGPRG (NP_001157849) VersaClone cDNA
Mas-related G Protein-coupled Receptor Member G
Brand:  R&D Systems™ RDC0727
Product Code. 18461450
  • 322.00 € / 10µg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
The Human MRGPRG (NP_001157849) VersaClone cDNA clone contains the complete ORF for MRGPRG, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MRGPRG insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hMRGPRG
10 μg at 0.2 μg/μL
882bp
Human
NP_001157849
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Reasearch Use Only; Not for Use in Humans