missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspergillus terrus Blasticidin S Deaminase, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Aspergillus terrus Blasticidin S Deaminase.
Brand: Abnova PAB7931.100uL
This item is not returnable.
View return policy
Specifications
| Aspergillus Terrus Blasticidin S Deaminase | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Aspergillus Terrus Blasticidin S Deaminase. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of Aspergillus Terrus Blasticidin S Deaminase. | |
| RUO | |
| Fungi | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction