Learn More
R&D Systems™ Polystyrene Microplates
high-binding, flat-bottom polystyrene microplates
Brand: R&D Systems™ DY990
This item is not returnable.
View return policy
Description
Specifications
Standard | |
Clear | |
Polystyrene | |
96 | |
ELISA |
8 x 12 | |
ELISA Microplate | |
360 μL | |
Flat Bottom (F-well) |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Polystyrene Microplates > Clear
Spot an opportunity for improvement?Share a Content Correction