Learn More
Velp Scientifica™ JP Recirculating Water Pump
Ensures optimum aspiration depending on the digestion phases and the number of samples placed in the digester connected to the pump. Velp Scientifica™ JP Recirculating Water Pumps eliminate the problem of unreliable water supply.
1350.00 €
Specifications
Product Type Recirculating Water Pump
Voltage 230 V
Material ABS
View More Specs
Products 1
Product Code Brand Hertz Price Quantity & Availability  
Product Code Brand Hertz Price Quantity & Availability  
11751164
View Documents Promotion Details
Velp Scientifica™
F30620198
50 Hz This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Aspiration matches digestion phases and sample number
  • Up to 35 liters/minute airflow pumping speed
  • Selectable performance mode related to samples
  • Chemically resistant ABS structure
  • Premium quality materials
  • Water recirculated for economy
  • Quiet operation
  • Water level detection
  • Convenient drain valve
  • Easy to move using two inset handles
Specifications
Specifications
Recirculating Water Pump
230 V
ABS