missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Vitamin B12-binding Protein
A DNA sequence encoding the Escherichia coli (strain K12) Vitamin B12-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8779-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_414700.1 | |
Vitamin B12-binding Protein | |
Research Use Only | |
btuF | |
Recombinant Protein | |
E. coli | |
PRVITLSPAN TELAFAAGIT PVGVSSYSDY PPQAQKIEQV STWQGMNLER IVALKPDLVI AWRGGNAERQ VDQLASLGIK VMWVDATSIE QIANALRQLA PWSPQPDKAE QAAQSLLDQY AQLKAQYADK PKKRVFLQFG INPPFTSGKE SIQNQVLEVC GGENIFKDSR VPWPQVSREQ VLARSPQAIV ITGGPDQIPK IKQYWGEQLK IPVIPLTSDW FERASPRIIL AAQQLCNALS QVD | |
Greater than 90% as determined by SDS-PAGE. |
947574 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |