missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant F. graminearum Galactose Oxidase His Protein
Brand: R&D Systems™ 11206-GX-050
This item is not returnable.
View return policy
Description
Measured by its ability to oxidize galactose. The specific activity is >75,000 pmol/min/μg, as measured under the described conditions.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
I1S2N3.1 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. | |
Unconjugated | |
Galactose Oxidase |
Enzyme Activity | |
M.W. Observed: 60-68 kDa, under reducing conditions., M.W. Theoretical: 70 kDa | |
E. coli-derived f. graminearum Galactose Oxidase protein Ala42-Gln680, with a C-terminal 6-His tag | |
<1.0 EU per 1 μg of the protein by the LAL method. | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction