missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-1b Biotin Protein
A cDNA sequence encoding the HCV Core Genotype-1b Biotin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12147-1mg
This item is not returnable.
View return policy
Specifications
Hepatitis C HCV Core Genotype-1b Biotin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-HCl pH 8 and 8M urea. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
Protein is >95% pure as determined by SDS-PAGE. |