missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AGO2 / Argonaute 2 / EIF2C2 Protein
A DNA sequence encoding the Homo sapiens (Human) AGO2 / Argonaute 2 / EIF2C2, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001158095.1 |
---|---|
Gene ID (Entrez) | 27161 |
Name | AGO2 / Argonaute 2 / EIF2C2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093511
|
enQuireBio™
QP8243-ye-10ug |
10 μg | N/A | ||||||
16033521
|
enQuireBio™
QP8243-ye-50ug |
50 μg | N/A | ||||||
16083511
|
enQuireBio™
QP8243-ye-100ug |
100 μg |
672.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013521
|
enQuireBio™
QP8243-ye-200ug |
200 μg | N/A | ||||||
16023521
|
enQuireBio™
QP8243-ye-500ug |
500 μg |
1810.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16003521
|
enQuireBio™
QP8243-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_001158095.1 | |
AGO2 / Argonaute 2 / EIF2C2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
27161 | |
Research Use Only | |
AGO2 | |
Recombinant Protein | |
Yeast | |
LVVVILPGKT PVYAEVKRVG DTVLGMATQC VQMKNVQRTT PQTLSNLCLK INVKLGGVNN ILLPQGRPPV FQQPVIFLGA DVTHPPAGDG KKPSIAAVVG SMDAHPNRYC ATVRVQQHRQ EIIQDLAAMV RELLIQFYKS TRFKPTRIIF YRDGVSEGQF QQVLHHELLA IREACIKLEK DYQPGITFIV VQKRHHTRLF CTDKNERVGK SGNIPAGTTV DTKITHPTEF DFYLCSHAGI QGTSRPSHYH VLWDDNRFSS DELQILTYQL CHTYVRCTRS VSIPAPAYYA HLVAFRARYH LV | |
Greater than 80% as determined by SDS-PAGE. |