missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Appetite-regulating hormone Protein
A DNA sequence encoding the Homo sapiens (Human) Appetite-regulating hormone, was expressed in the hosts and tags indicated.
160.00 € - 2485.00 €
Specifications
Accession Number | NP_001128413.1 |
---|---|
Gene ID (Entrez) | 51738 |
Name | Appetite-regulating hormone Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16092671
|
enQuireBio™
QP8104-ec-10ug |
10 μg | N/A | ||||||
16032681
|
enQuireBio™
QP8104-ec-50ug |
50 μg | N/A | ||||||
16082671
|
enQuireBio™
QP8104-ec-100ug |
100 μg |
554.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16012681
|
enQuireBio™
QP8104-ec-200ug |
200 μg | N/A | ||||||
16022681
|
enQuireBio™
QP8104-ec-500ug |
500 μg | N/A | ||||||
16002681
|
enQuireBio™
QP8104-ec-1mg |
1 mg |
2485.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001128413.1 | |
Appetite-regulating hormone Protein | |
Not determined. | |
Full Length of BC025791 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
51738 | |
Research Use Only | |
GHRL | |
Recombinant Protein | |
E. coli | |
MPSPGTVCSL LLLGMLWLDL AMAGSSFLSP EHQRVQQRKE SKKPPAKLQP RALAGWLRPE DGGQAEGAED EMEVRFNAPF DVGIKLSGVQ YQQHSQALGK FLQDILWEEA KEAPADK | |
Greater than 80% as determined by SDS-PAGE. |