missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTB / POZ domain-containing protein KCTD15 Protein
A DNA sequence encoding the Homo sapiens (Human) BTB/POZ domain-containing protein KCTD15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7877-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001123466.1 | |
BTB / POZ domain-containing protein KCTD15 Protein | |
Research Use Only | |
KCTD15 | |
Recombinant Protein | |
E. coli | |
MPHRKERPSG SSLHTHGSTG TAEGGNMSRL SLTRSPVSPL AAQGIPLPAQ LTKSNAPVHI DVGGHMYTSS LATLTKYPDS RISRLFNGTE PIVLDSLKQH YFIDRDGEIF RYVLSFLRTS KLLLPDDFKD FSLLYEEARY YQLQPMVREL ERWQQEQEQR RRSRACDCLV VRVTPDLGER IALSGEKALI EEVFPETGDV MCNSVNAGWN QDPTHVIRFP LNGYCRLNSV QDVL | |
Greater than 80% as determined by SDS-PAGE. |
79047 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |