missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BTB / POZ domain-containing protein KCTD15 Protein

A DNA sequence encoding the Homo sapiens (Human) BTB/POZ domain-containing protein KCTD15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7877-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011341

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001123466.1
BTB / POZ domain-containing protein KCTD15 Protein
Research Use Only
KCTD15
Recombinant Protein
E. coli
MPHRKERPSG SSLHTHGSTG TAEGGNMSRL SLTRSPVSPL AAQGIPLPAQ LTKSNAPVHI DVGGHMYTSS LATLTKYPDS RISRLFNGTE PIVLDSLKQH YFIDRDGEIF RYVLSFLRTS KLLLPDDFKD FSLLYEEARY YQLQPMVREL ERWQQEQEQR RRSRACDCLV VRVTPDLGER IALSGEKALI EEVFPETGDV MCNSVNAGWN QDPTHVIRFP LNGYCRLNSV QDVL
Greater than 80% as determined by SDS-PAGE.
79047
500 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions