missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK4 Protein
A DNA sequence encoding the Homo sapiens (Human) CDK4, was expressed in the hosts and tags indicated.
580.00 € - 1600.00 €
Specifications
Name | CDK4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | CDK4 |
Protein Length | Extracellular Domain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952788
|
enQuireBio™
QP10035-ba-20ug |
20 μg |
580.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15962788
|
enQuireBio™
QP10035-ba-50ug |
50 μg | N/A | ||||||
15942788
|
enQuireBio™
QP10035-ba-100ug |
100 μg |
1600.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CDK4 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
CDK4 | |
Recombinant Protein | |
Baculovirus | |
MATSRYEPVA EIGVGAYGTV YKARDPHSGH FVALKSVRVP NGGGGGGGLP ISTVREVALL RRLEAFEHPN VVRLMDVCAT SRTDREIKRC FVESLSSVET LKPTSWAKSL T | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title