Learn More
enQuireBio™ Recombinant Human EEF1E1 / AIMP3 / p18 Protein
Product Code. 15938959
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 15938959
Brand: enQuireBio™ QP5961ec200ug
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A DNA sequence encoding the Homo sapiens (Human) EEF1E1 / AIMP3 / p18, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001129122.1
Gene ID (Entrez) 9521
Name EEF1E1 / AIMP3 / p18 Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol EEF1E1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence AAAAELSLLE KSLGLSKGNK YSAQGERQIP VLQTNNGPSL TGLTTIAAHL VKQANKEYLL GSTAEEKAIV QQWLEYRVTQ VDGHSSKNDI HTLLKDLNSY LEDKVYLTGY NFTLADILLY YGLHRFIVDL TVQEKEKYLN VSRWFCHIQH YPGIRQHLSS VVFIKNRLYT NSH
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.