missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human FGF-10 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-34927-5ug
This item is not returnable.
View return policy
Description
Fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 106IU/mg.
Specifications
O15520, O15520, O15520 | |
PAGE, Bioactivity | |
2255 | |
5 μg | |
Less than 1 EU/ug of FGF-10 as determined by LAL method. | |
FGF10 | |
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
FGF-10 |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4. | |
19.1 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF-2/FGF-10, produced by fibroblasts of urinary bladder lamina propria | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>97% pure by SDS-PAGE and HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human FGF-10 Protein > 5 μg
Spot an opportunity for improvement?Share a Content Correction