missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-17 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 319-FG-025
209.06 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human FGF-17 Protein is derived from E. coli. The Recombinant Human FGF-17 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
22.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. | |
Recombinant |
8822 | |
FGF-17 | |
E. coli-derived human FGF-17 protein Thr23-Thr216,with and without an N-terminal Met | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |