missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GADD45G / CR6 Protein

A DNA sequence encoding the Homo sapiens (Human) GADD45G / CR6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6064-ec-500ug

 View more versions of this product

Product Code. 15969719

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_006696.1
GADD45G / CR6 Protein
Research Use Only
GADD45G
Recombinant Protein
E. coli
MTLEEVRGQD TVPESTARMQ GAGKALHELL LSAQRQGCLT AGVYESAKVL NVDPDNVTFC VLAAGEEDEG DIALQIHFTL IQAFCCENDI DIVRVGDVQR LAAIVGAGEE AGAPGDLHCI LISNPNEDAW KDPALEKLSL FCEESRSVND WVPSITLPE
Greater than 90% as determined by SDS-PAGE.
10912
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions