missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HOXA9 Protein

A cDNA sequence encoding the HOXA9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12295-1mg

 View more versions of this product

Product Code. 15939968

  • 4090.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HOXA9 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMATTGAL GNYYVDSFLL GADAADELSV GRYAPGTLGQ PPRQAATLAE HPDFSPCSFQ SKATVFGASW NPVHAAGANA VPAAVYHHHH HHPYVHPQAP VAAAAPDGRY MRSWLEPTPG ALSFAGLPSS RPYGIKPEPL SARRGDCPTL DTHTLSLTDY ACGSPPVDRE KQPSEGAFSE NNAENESGGD KPPIDPNNPA ANWLHARSTR KKRCPYTKHQ TLELEKEFLF NMYLTRDRRY EVARLLNLTE RQVKIWFQNR RMKMKKINKD RAKDE
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HOXA9 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions