missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IDI2 Protein
A cDNA sequence encoding the IDI2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12367-1mg
This item is not returnable.
View return policy
Specifications
91734 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSDINLDWVD RRQLQRLEEM LIVVDENDKV IGADTKRNCH LNENIEKGLL HRAFSVVLFN TKNRILIQQR SDTKVTFPGY FTDSCSSHPL YNPAELEEKD AIGVRRAAQR RLQAELGIPG EQISPEDIVF MTIYHHKAKS DRIWGEHEIC YLLLVRKNVT LNPDPSETKS ILYLSQEELW ELLEREARGE VKVTPWLRTI AERFLYRWWP HLDDVTPFVE LHKIHRV | |
Greater than 80.0% as determined by SDS-PAGE. |
IDI2 Protein | |
Research Use Only | |
IDI2 | |
Human | |
His | |
IDI2 solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1mM PMSF. |