missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIR2DS4 Protein
A cDNA sequence encoding the KIR2DS4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12491-5ug
This item is not returnable.
View return policy
Specifications
KIR2DS4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEGVHRKPSF LALPGHLVKS EETVILQCWS DVMFEHFLLH REGKFNNTLH LIGEHHDGVS KANFSIGPMM PVLAGTYRCY GSVPHSPYQL SAPSDPLDMV IIGLYEKPSL SAQPGPTVQA GENVTLSCSS RSSYDMYHLS REGEAHERRL PAVRSINGTF QADFPLGPAT HGGTYRCFGS FRDAPYEWSN SSDPLLVSVT GN | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
KIR2DS4 protein solution (1 mg/ml) contains 20mM Tris-HCl (pH 7.5). |