missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LAMP3 Protein

A cDNA sequence encoding the LAMP3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12532-10ug

 View more versions of this product

Product Code. 15950729

  • 222.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

LAMP3 Protein
Research Use Only
Recombinant Protein
Baculovirus
KAFPETRDYS QPTAAATVQD IKKPVQQPAK QAPHQTLAAR FMDGHITFQT AATVKIPTTT PATTKNTATT SPITYTLVTT QATPNNSHTA PPVTEVTVGP SLAPYSLPPT ITPPAHTTGT SSSTVSHTTG NTTQPSNQTT LPATLSIALH KSTTGQKPVQ PTHAPGTTAA AHNTTRTAAP ASTVPGPTLA PQPSSVKTGI YQVLNGSRLC IKAEMGIQLI VQDKESVFSP RRYFNIDPNA TQASGNCGTR KSNLLLNFQG GFVNLTFTKD EESYYISEVG AYLTVSDPET IYQGIKHAVV MFQTAVGHSF KCVSEQSLQL SAHLQVKTTD VQLQAFDFED DHFGNVDECS SDYTLEHHHH HH
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
LAMP3 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions