missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LTF Holo Protein
A cDNA sequence encoding the LTF Holo was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12596-100mg
This item is not returnable.
View return policy
Specifications
4057 | |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Rice | |
MKLVFLVLLF LGALGLCLAG RRRSVQWCAV SQPEATKCFQ WQRNMRKVRG PPVSCIKRDS PIQCIQAIAE NRADAVTLDG GFIYEAGLAP YKLRPVAAEV YGTERQPRTH YYAVAVVKKG GSFQLNELQG LKSCHTGLRR TAGWNVPIGT LRPFLNWTGP PEPIEAAVAR FFSASCVPGA DKGQFPNLCR LCAGTGENKC AFSSQEPYFS YSGAFKCLRD GAGDVAFIRE STVFEDLSDE AERDEYELLC PDNTRKPVDK FKDCHLARVP SHAVVARSVN GKEDAIWNLL RQAQEKFGKD KSPKFQLFGS PSGQKDLLFK DSAIGFSRVP PRIDSGLYLG SGYFTAIQNL RKSEEEVAAR RARVVWCAVG EQELRKCNQW SGLSEGSVTC SSASTTEDCI ALVLKGEADA MSLDGGYVYT AGKCGLVPVL AENYKSQQSS DPDPNCVDRP VEGYLAVAVV RRSDTSLTWN SVKGKKSCHT AVDRTAGWNI PMGLLFNQTG SCKFDEYFSQ SCAPGSDPRS NLCALCIGDE QGENKCVPNS NERYYGYTGA FRCLAENAGD VAFVKDVTVL QNTDGNNNEA WAKDLKLADF ALLCLDGKRK PVTEARSCHL AMAPNHAVVS RMDKVERLKQ VLLHQQAKFG RNGSDCPDKF CLFQSETKNL LFNDNTECLA RLHGKTTYEK YLGPQYVAGI TNLKKCSTSP LLEACEFLRK | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
LTF Holo Protein | |
Research Use Only | |
LTF | |
Human | |
Untagged | |
The Human Holo lactotransferrin was lyophilized from water, contains 1.66 mg iron/gram Lactoferrin. |