missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MMP-3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 513-MP-010
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >150 pmol/min/μg, as measured under the described conditions.Specifications
P08254 | |
Supplied as a 0.2 μm filtered solution in MES, NaCl, CaCl2, Glycerol and Brij-35. | |
M.W. (Observed): 54-56 kDa doublet, reducing conditions; M.W. (theoretical): 52 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Enzyme Activity | |
4314 | |
MMP-3 | |
Mouse myeloma cell line, NS0-derived human MMP-3 protein Tyr18-Cys477 (Lys45Glu) | |
Pro form | |
MMP3 | |
Recombinant | |
MMP-3 |