missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MUCL1 Protein
A cDNA sequence encoding the MUCL1 was constructed and used to recombinantly synthesize the protein.
137.00 € - 1815.00 €
Specifications
Gene ID (Entrez) | 118430 |
---|---|
Name | MUCL1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | MUCL1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961429
|
enQuireBio™
QP12751-2ug |
2 μg |
137.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951429
|
enQuireBio™
QP12751-10ug |
10 μg |
212.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941429
|
enQuireBio™
QP12751-100ug |
100 μg |
1815.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
118430 | |
Research Use Only | |
MUCL1 | |
Human | |
His | |
MUCL1 protein solution (0.25 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol. |
MUCL1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSNPTTAAP ADTYPATGPA DDEAPDAETT AAATTATTAA PTTATTAAST TARKDIPVLP KWVGDLPNGR VCP | |
Greater than 85.0% as determined by SDS-PAGE. |