missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NCR2 Protein

A cDNA sequence encoding the NCR2 was constructed and used to recombinantly synthesize the protein.

137.00 € - 4090.00 €

Specifications

Name NCR2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15971589
View Documents Promotion Details
enQuireBio™
QP12803-5ug
5 μg
137.00 €
5µg
Please to purchase this item. Need a web account? Register with us today!
15961589
View Documents Promotion Details
enQuireBio™
QP12803-20ug
20 μg
212.00 €
20µg
Please to purchase this item. Need a web account? Register with us today!
15951589
View Documents Promotion Details
enQuireBio™
QP12803-1mg
1 mg
4090.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NCR2 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NCR2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSQAQSKAQV LQSVAGQTLT VRCQYPPTGS LYEKKGWCKE ASALVCIRLV TSSKPRTMAW TSRFTIWDDP DAGFFTVTMT DLREEDSGHY WCRIYRPSDN SVSKSVRFYL VVS
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications