missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFAF4 Protein
A cDNA sequence encoding the NDUFAF4 was constructed and used to recombinantly synthesize the protein.
137.00 € - 1815.00 €
Specifications
Name | NDUFAF4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931639
|
enQuireBio™
QP12815-1ug |
1 μg |
137.00 €
1µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951639
|
enQuireBio™
QP12815-5ug |
5 μg |
212.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941639
|
enQuireBio™
QP12815-50ug |
50 μg |
1815.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NDUFAF4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NDUFAF4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGALVIR GIRNFNLENR AEREISKMKP SVAPRHPSTN SLLREQISLY PEVKGEIARK DEKLLSFLKD VYVDSKDPVS SLQVKAAETC QEPKEFRLPK DHHFDMINIK SIPKGKISIV EALTLLNNHK LFPETWTAEK IMQEYQLEQK DVNSLLKYFV TFEVEIFPPE DKKAIRSK | |
Greater than 85.0% as determined by SDS-PAGE. |