missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OGT1 Protein
A DNA sequence encoding the Homo sapiens (Human) OGT1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6450-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_858058.1 | |
OGT1 Protein | |
Research Use Only | |
OGT | |
Recombinant Protein | |
E. coli | |
MAEANHFIDL SQIPCNGKAA DRIHQDGIHI LVNMNGYTKG ARNELFALRP APIQAMWLGY PGTSGALFMD YIITDQETSP AEVAEQYSEK LAYMPHTFFI GDHANMFPHL KKKAVIDFKS NGHIYDNRIV LNGIDLKAFL DSLPDVKIVK MKCPDGGDNA DSSNTALNMP VIPMNTIAEA VIEMINRGQI QITINGFSIS NGLATTQINN KAATGEEVPR TIIVTTRSQY GLPEDAIVYC NFNQLYKIDP STLQMWANIL KRVPNSVLWL LRFPAVGEPN IQQYAQNMGL PQNRIIFSPV APKEEHVRRG QLADVCLDTP LCNGHTTGMD VLWAGTPMVT MPGETLASRV AASQLTCLGC LELIAKNRQE YEDIAVKLGT DLEYLKKVRG KVWKQRISSP LFNTKQYTME LERLYLQ | |
Greater than 90% as determined by SDS-PAGE. |
8473 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |