missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAPP A Protein
A cDNA sequence encoding the PAPP A was constructed and used to recombinantly synthesize the protein.
137.00 € - 7270.00 €
Specifications
Name | PAPP A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15972069
|
enQuireBio™
QP12957-2ug |
2 μg |
137.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952069
|
enQuireBio™
QP12957-10ug |
10 μg |
212.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15962069
|
enQuireBio™
QP12957-1mg |
1 mg |
7270.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PAPP A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PAPP-A protein is supplied in 1x PBS and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRLWSWVLHL GLLSAALGCG LAERPRRARR DPRAGRPPRP AAGPATCATR AARGRRASPP PPPPPGGAWE AVRVPRRRQQ REARGATEEP SPPSRALYFS GRGEQLRLRA DLELPRDAFT LQVWLRAEGG QRSPAVITGL YDKCSYISRD RGWVVGIHTI SDQDNKDPRY FFSLKTDRAR QVTTINAHRS YLPGQWVYLA ATYDGQFMKL YVNGAQVATS GEQVGGIFSP LTQKCKVLML GGSALNHNYR GYIEHFSLWK VARTQREILS DMETHGAHTA LPQLLLQENW DNVKHAWSPM KDGSSPKVEF SNAHGFLLDT SLEPPLCGQT LCDNTEVIAS YNQLSSFRQP KVVRYRVVNL YEDDHKNPTV TREQVDFQHH QLAEAFKQYN ISWELDVLEV SNSSLRRRLI LANCDISKIG DENCDPECNH TLTGHDGGDC RHLRHPAFVK KQHNGVCDMD CNYERFNFDG GECCDPEITN VTQTCFDPDS PHRAYLDVNE LKNILKLDGS THLNIFFAKS SEEELAGVAT WPWDKEALMH LGGIVLNPSF YGMPGHTHTM IHEIGHSLGL YHVFRGISEI QSCSDPCMET EPSFETGDLC NDTNPAPKHK SCGDPGPGND TCGFHSFFNT PYNNFMSYAD DDCTDSFTPN QVARMHCYLD LVYQGWQPSR KPAPVALAPQ VLGHTTDSVT LEWFPPIDGH FFERELGSAC HLCLEGRILV QYASNASSPM PCSPSGHWSP REAEGHPDVE QPCKSSVRTW SPNSAVNPHT VPPACPEPQG CYLELEFLYP LVPESLTIWV TFVSTDWDSS GAVNDIKLLA VSGKNISLGP QNVFCDVPLT IRLWDVGEEV YGIQIYTLDE HLEIDAAMLT STADTPLCLQ CKPLKYKVVR DPPLQMDVAS ILHLNRKFVD | |
Greater than 95.0% as determined by SDS-PAGE. |