missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDCL3 Protein
A cDNA sequence encoding the PDCL3 was constructed and used to recombinantly synthesize the protein.
143.00 € - 5670.00 €
Specifications
Name | PDCL3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962169
|
enQuireBio™
QP12989-5ug |
5 μg |
143.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952169
|
enQuireBio™
QP12989-20ug |
20 μg |
222.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942169
|
enQuireBio™
QP12989-1mg |
1 mg |
5670.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PDCL3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PDCL3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQDPNAD TEWNDILRKK GILPPKESLK ELEEEAEEEQ RILQQSVVKT YEDMTLEELE DHEDEFNEED ERAIEMYRRR RLAEWKATKL KNKFGEVLEI SGKDYVQEVT KAGEGLWVIL HLYKQGIPLC ALINQHLSGL ARKFPDVKFI KAISTTCIPN YPDRNLPTIF VYLEGDIKAQ FIGPLVFGGM NLTRDELEWK LSESGAIMTD LEENPKKPIE DVLLSSVRRS VLMKRDSDSE GD | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title