missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDIA6 Protein
A cDNA sequence encoding the PDIA6 was constructed and used to recombinantly synthesize the protein.
137.00 € - 212.00 €
Specifications
Gene ID (Entrez) | 10130 |
---|---|
Name | PDIA6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | PDIA6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962189
|
enQuireBio™
QP12997-5ug |
5 μg |
137.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952189
|
enQuireBio™
QP12997-20ug |
20 μg |
212.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
10130 | |
Research Use Only | |
PDIA6 | |
Human | |
His | |
The PDIA6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT and 50mM NaCl. |
PDIA6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLYSSSDDVI ELTPSNFNRE VIQSDSLWLV EFYAPWCGHC QRLTPEWKKA ATALKDVVKV GAVDADKHHS LGGQYGVQGF PTIKIFGSNK NRPEDYQGGR TGEAIVDAAL SALRQLVKDR LGGRSGGYSS GKQGRSDSSS KKDVIELTDD SFDKNVLDSE DVWMVEFYAP WCGHCKNLEP EWAAAASEVK EQTKGKVKLA AVDATVNQVL ASRYGIRGFP TIKIFQKGES PVDYDGGRTR SDIVSRALDL FSDNAPPPEL LEIINEDIAK RTCEEHQLCV VAVLPHILDT GAAGRNSYLE VLLKLADKYK KKMWGWLWTE AGAQSELETA LGIGGFGYPA MAAINARKMK FALLKGSFSE QGINEFLREL SFGRGSTAPV GGGAFPTIVE REPWDGRDGE LPVEDDIDLS DVELDDLGKD EL | |
Greater than 90.0% as determined by SDS-PAGE. |