missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA2G7 Protein
A cDNA sequence encoding the PLA2G7 was constructed and used to recombinantly synthesize the protein.
137.00 € - 4090.00 €
Specifications
Name | PLA2G7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952429
|
enQuireBio™
QP13074-HIS-HEK-5ug |
5 μg |
137.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942429
|
enQuireBio™
QP13074-HIS-HEK-20ug |
20 μg |
212.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15932429
|
enQuireBio™
QP13074-HIS-HEK-1mg |
1 mg |
4090.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PLA2G7 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PLA2G7 protein is supplied in 20mM Tris-HCl pH 8.0, 1mM EDTA and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
MVPPKLHVLF CLCGCLAVVY PFDWQYINPV AHMKSSAWVN KIQVLMAAAS FGQTKIPRGN GPYSVGCTDL MFDHTNKGTF LRLYYPSQDN DRLDTLWIPN KEYFWGLSKF LGTHWLMGNI LRLLFGSMTT PANWNSPLRP GEKYPLVVFS HGLGAFRTLY SAIGIDLASH GFIVAAVEHR DRSASATYYF KDQSAAEIGD KSWLYLRTLK QEEETHIRNE QVRQRAKECS QALSLILDID HGKPVKNALD LKFDMEQLKD SIDREKIAVI GHSFGGATVI QTLSEDQRFR CGIALDAWMF PLGDEVYSRI PQPLFFINSE YFQYPANIIK MKKCYSPDKE RKMITIRGSV HQNFADFTFA TGKIIGHMLK LKGDIDSNVA IDLSNKASLA FLQKHLGLHK DFDQWDCLIE GDDENLIPGT NINTTNQHIM LQNSSGIEKY N | |
Greater than 95.0% as determined by SDS-PAGE. |