missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLB Protein
A cDNA sequence encoding the POLB was constructed and used to recombinantly synthesize the protein.
137.00 € - 4090.00 €
Specifications
Gene ID (Entrez) | 5423 |
---|---|
Name | POLB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | POLB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962479
|
enQuireBio™
QP13091-5ug |
5 μg |
137.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952479
|
enQuireBio™
QP13091-20ug |
20 μg |
212.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942479
|
enQuireBio™
QP13091-1mg |
1 mg |
4090.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
5423 | |
Research Use Only | |
POLB | |
Human | |
His | |
POLB protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl. |
POLB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSKRKAPQET LNGGITDMLT ELANFEKNVS QAIHKYNAYR KAASVIAKYP HKIKSGAEAK KLPGVGTKIA EKIDEFLATG KLRKLEKIRQ DDTSSSINFL TRVSGIGPSA ARKFVDEGIK TLEDLRKNED KLNHHQRIGL KYFGDFEKRI PREEMLQMQD IVLNEVKKVD SEYIATVCGS FRRGAESSGD MDVLLTHPSF TSESTKQPKL LHQVVEQLQK VHFITDTLSK GETKFMGVCQ LPSKNDEKEY PHRRIDIRLI PKDQYYCGVL YFTGSDIFNK NMRAHALEKG FTINEYTIRP LGVTGVAGEP LPVDSEKDIF DYIQWKYREP KDRSE | |
Greater than 90.0% as determined by SDS-PAGE. |