missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMA6 Protein
A cDNA sequence encoding the PSMA6 was constructed and used to recombinantly synthesize the protein.
137.00 € - 7875.00 €
Specifications
Name | PSMA6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952769
|
enQuireBio™
QP13185-2ug |
2 μg |
137.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15932769
|
enQuireBio™
QP13185-10ug |
10 μg |
212.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942769
|
enQuireBio™
QP13185-1mg |
1 mg |
7875.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PSMA6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PSMA6 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 40% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSRGSS AGFDRHITIF SPEGRLYQVE YAFKAINQGG LTSVAVRGKD CAVIVTQKKV PDKLLDSSTVTHLFKITENI GCVMTGMTAD SRSQVQRARY EAANWKYKYG YEIPVDMLCK RIADISQVYT QNAEMRPLGC CMILIGIDEE QGPQVYKCDP AGYYCGFKAT AAGVKQTEST SFLEKKVKKK FDWTFEQTVE TAITCLSTVL SIDFKPSEIE VGVVTVENPK FRILTEAEID AHLVALAERD | |
Greater than 90.0% as determined by SDS-PAGE. |