missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB10 Protein
A cDNA sequence encoding the RAB10 was constructed and used to recombinantly synthesize the protein.
137.00 € - 7875.00 €
Specifications
Name | RAB10 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952909
|
enQuireBio™
QP13228-2ug |
2 μg |
137.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15932909
|
enQuireBio™
QP13228-10ug |
10 μg |
212.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942909
|
enQuireBio™
QP13228-1mg |
1 mg |
7875.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
RAB10 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB10 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAKKTYDLLF KLLLIGDSGV GKTCVLFRFS DDAFNTTFIS TIGIDFKIKT VELQGKKIKL QIWDTAGQER FHTITTSYYR GAMGIMLVYD ITNGKSFENI SKWLRNIDEH ANEDVERMLL GNKCDMDDKR VVPKGKGEQI AREHGIRFFE TSAKANINIE KAFLTLAEDI LRKTPVKEPN SENVDISSGG GVTGWKSKCC | |
Greater than 90% as determined by SDS-PAGE. |