missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNLS Protein

A cDNA sequence encoding the RNLS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13333-1mg

 View more versions of this product

Product Code. 15953239

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

RNLS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMALLRRQ TSGPLYLAVW DKAEDSGGRM TTACSPHNPQ CTADLGAQYI TCTPHYAKKH QRFYDELLAY GVLRPLSSPI EGMVMKEGDC NFVAPQGISS IIKHYLKESG AEVYFRHRVT QINLRDDKWE VSKQTGSPEQ FDLIVLTMPV PEILQLQGDI TTLISECQRQ QLEAVSYSSR YALGLFYEAG TKIDVPWAGQ YITSNPCIRF VSIDNKKRNI ESSEIGPSLV IHTTVPFGVT YLEHSIEDVQ ELVFQQLENI LPGLPQPIAT KCQKWRHSQV TNAAANCPGQ MTLHHKPFLA CGGDGFTQSN FDGCITSALC VLEALKNYI
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RNLS solution contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions