missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIRT1 / SIR2L1 Protein
A DNA sequence encoding the Homo sapiens (Human) SIRT1 / SIR2L1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1351-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001135970.1 | |
SIRT1 / SIR2L1 Protein | |
Research Use Only | |
SIRT1 | |
Recombinant Protein | |
E. coli | |
ADEAALALQP GGSPSAAGAD REAASSPAGE PLRKRPRRDG PGLERSPGEP GGAAPEREVP AAARGCPGAA AAALWREAEA EAAAAGGEQE AQATAAAGEG DNGPGLQGPS REPPLADNLY DEDDDDEGEE EEEAAAAAIG YRDNLLFGDE IITNGFHSCE SDEEDRASHA SSSDWTPRPR IGPYTFVQQH LMIGTDPRTI LKDLLPETIP PPELDDMTLW QIVINILSEP PKRKKRKDIN TIEDAVKLLQ ECKKIIVLTG AGVSVSCGIP DFRSRDGIYA RLAVDFPDLP DPQAMFDIEY FRKDPRPFFK FAKEIYPGQF QPSLCHKFIA LSDKEGKLLR NYTQNIDTLE QVAGIQRIIQ CHGSFATASC LICKYKVDCE AVRGDIFNQV VPRCPRCPAD EPLAIMKPEI VFFGENLPEQ FHRAMKYDKD EVDLLIVIGS SLKVRPVALI PSSIPHEVPQ ILINREPLPH LHFDVELLGD CDVIINELCH RLGGEYAKLC CNPVKLSEIT EKPPRTQKEL AYLSELPPTP LHVSEDSSSP ERTSPPDSSV IVTLLDQAAK SNDDLDVSES KGCMEEKPQE VQTSRNVESI AEQMENPDLK NVGSSTGEKN ERTSVAGTVR KCWPNRVAKE QISRRLDGNQ YLFLPPNRYI FHGAEVYSDS EDDVLSSSSC GSNSDSGTCQ SPSLEEPMED ESEIEEFYNG LEDEPDVPER AGGAGFGTDG DDQEAINEAI SVKQEVTDMN YPSNKS | |
Greater than 90% as determined by SDS-PAGE. |
23411 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |