missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human SOST/Sclerostin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
508.00 €
Specifications
Gene ID (Entrez) | 50964 |
---|---|
Molecular Weight (g/mol) | 28-32 kDa, reducing conditions |
Name | SOST/Sclerostin |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived human SOST/Sclerostin protein Gln24-Tyr213,with an N-terminal 7-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18117710
|
R&D Systems™
1406-ST-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
508.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18118600
|
R&D Systems™
1406-ST-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
508.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human SOST/Sclerostin Protein is derived from NS0. The Recombinant Human SOST/Sclerostin Protein has been validated for the following applications: Bioactivity.Specifications
50964 | |
SOST/Sclerostin | |
Mouse myeloma cell line,NS0-derived human SOST/Sclerostin protein Gln24-Tyr213,with an N-terminal 7-His tag | |
Monomer | |
Recombinant |
28-32 kDa, reducing conditions | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |