missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX18 Protein
A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6766-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001073977.1 | |
T-box transcription factor TBX18 Protein | |
Research Use Only | |
TBX18 | |
Recombinant Protein | |
E. coli | |
MAEKRRGSPC SMLSLKAHAF SVEALIGAEK QQQLQKKRRK LGAEEAAGAV DDGGCSRGGG AGEKGSSEGD EGAALPPPAG ATSGPARSGA DLERGAAGGC EDGFQQGASP LASPGGSPKG SPARSLARPG TPLPSPQAPR VDLQGAELWK RFHEIGTEMI ITKAGRRMFP AMRVKISGLD PHQQYYIAMD IVPVDNKRYR YVYHSSKWMV AGNADSPVPP RVYIHPDSPA SGETWMRQVI SFDKLKLTNN ELDDQGHIIL HSMHKYQPRV HVIRKDCGDD LSPIKPVPSG EGVKAFSFPE TVFTTVTAYQ NQQITRLKID RNPFAKGFRD SGRNRMGLEA LVESYAFWRP SLRTLTFEDI PGIPKQGNAS SSTLLQGTGN GVPATHPHLL SGSSCSSPAF HLGPNTSQLC SLAPADYSAC ARSGLTLNRY STSLAETYNR LTNQAGETFA PPRTPSYVGV SSSTSVNMSM GGTDGDTFSC PQTSLSMQIS GMSPQLQYIM PSPSSNAFAT NQTHQGSYNT FRLHSPCALY GYNFSTSPKL AASPEKIVSS QGSFLGSSPS GTMTDRQMLP PVEGVHLLSS GGQQSFFDSR TLGSLTLSSS QVSAHMV | |
Greater than 90% as determined by SDS-PAGE. |
9096 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human T-box transcription factor TBX18 Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction